Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
Brand:  DWK Life Sciences A620030001211
 View more versions of this product
Product Code. 10676423
  • 42.85 € / Pack of 50
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Disposable
  • Borosilicate glass
  • Excellent resistance to temperature changes
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round bottom
50Pack
Test Tube
200 mm
30 mm