missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenoxy)-N,N-dimethylpropylamine, 97%, Thermo Scientific™

156.00 € - 481.00 €

Chemical Identifiers

CAS 912569-57-4
Molecular Formula C11H16BrNO
Molecular Weight (g/mol) 258.159
MDL Number MFCD03718729
InChI Key FMYWALTVPROHIA-UHFFFAOYSA-N
Synonym 3-3-bromophenoxy-n,n-dimethylpropylamine, 3-3-bromophenoxy-n,n-dimethylpropan-1-amine, 3-3-bromophenoxy propyl dimethylamine
PubChem CID 2183561
IUPAC Name 3-(3-bromophenoxy)-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCOC1=CC(=CC=C1)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10728845
View Documents
Thermo Scientific Maybridge
CC59410DA
1 g Amber Glass Bottle
156.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10402844
View Documents
Thermo Scientific Maybridge
CC59410DE
5 g Amber Glass Bottle
481.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

912569-57-4
258.159
FMYWALTVPROHIA-UHFFFAOYSA-N
2183561
CN(C)CCCOC1=CC(=CC=C1)Br
C11H16BrNO
MFCD03718729
3-3-bromophenoxy-n,n-dimethylpropylamine, 3-3-bromophenoxy-n,n-dimethylpropan-1-amine, 3-3-bromophenoxy propyl dimethylamine
3-(3-bromophenoxy)-N,N-dimethylpropan-1-amine

Specifications

912569-57-4
100.0
C11H16BrNO
3-3-bromophenoxy-n,n-dimethylpropylamine, 3-3-bromophenoxy-n,n-dimethylpropan-1-amine, 3-3-bromophenoxy propyl dimethylamine
CN(C)CCCOC1=CC(=CC=C1)Br
258.159
258.16
3-(3-Bromophenoxy)-N, N-dimethylpropylamine
96.5
97%
MFCD03718729
FMYWALTVPROHIA-UHFFFAOYSA-N
3-(3-bromophenoxy)-N,N-dimethylpropan-1-amine
2183561
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications