missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-Chlorophenyl)-3-oxopropanenitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | ClC6H4C(O)CH2CN |
Quantity | 1 g |
Formula Weight | 179.61 |
Percent Purity | 97% |
Chemical Name or Material | 3-(3-Chlorophenyl)-3-oxopropanenitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'