missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Di-n-heptyloxacarbocyanine iodide, 97%, Thermo Scientific™

Product Code. 15492128
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53213-83-5
Molecular Formula C31H41IN2O2
Molecular Weight (g/mol) 600.59
MDL Number MFCD00274413,MFCD00274413,MFCD00274413
InChI Key ANJZLSIIEGUCQL-UHFFFAOYSA-M
Synonym 3,3'-diheptyloxacarbocyanine iodide
PubChem CID 102600931
IUPAC Name (2E)-3-heptyl-2-[(Z)-3-(3-heptyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;iodide
SMILES [I-].CCCCCCCN1C(OC2=CC=CC=C12)=CC=CC1=[N+](CCCCCCC)C2=CC=CC=C2O1

Specifications

Color Red
Quantity 500 mg
Sensitivity Light sensitive
Formula Weight 600.58
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,3'-Di-n-heptyloxacarbocyanine iodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'