missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%, Thermo Scientific Chemicals

Product Code. 11401550
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101203-31-0
Molecular Formula C24H34O2
Molecular Weight (g/mol) 354.534
MDL Number MFCD01862439
InChI Key NMVVBVMYPLMIOU-UHFFFAOYSA-N
Synonym 3,3'-di-tert-butyl-5,5',6,6'-tetramethyl-1,1'-biphenyl-2,2'-diol, 5,5',6,6'-tetramethyl-3,3'-di-tert-butyl-1,1'-biphenyl-2,2'-diol, 3,3'-di-t-butyl-5,5',6,6'-tetramethyl-1,1'-biphenyl-2,2'-diol, s---3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 6-tert-butyl-2-3-tert-butyl-2-hydroxy-5,6-dimethylphenyl-3,4-dimethylphenol, 2,2'-bi 6-tert-butyl-3,4-dimethylphenol, r-3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, s-3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, as-3,3'-di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, s--5,5,6,6-tetramethyl-3,3-di-tert-butyl-1,1-biphenyl-2,2-diol
PubChem CID 3255344
IUPAC Name 6-tert-butyl-2-(3-tert-butyl-2-hydroxy-5,6-dimethylphenyl)-3,4-dimethylphenol
SMILES CC1=CC(=C(C(=C1C)C2=C(C(=CC(=C2O)C(C)(C)C)C)C)O)C(C)(C)C

Specifications

Melting Point 164°C to 169°C
Quantity 1 g
Formula Weight 354.54
Percent Purity 99%
Chemical Name or Material (+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'