missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, Thermo Scientific Chemicals

Product Code. 15444629
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
10 g

3,3'-Diaminobenzidine tetrahydrochloride hydrate, CAS # 868272-85-9, is a peroxidase substrate that is commonly used for immunoblotting and immunohistochemical staining techniques.

3,3?-diaminobenzidine-tetrahydrochloride-dihydrate (DAB) was used as a chromogen, for detecting the antigen-antibody complex. It is one of the most potent marker being used in immunochemistry, mostly at the electron and light microscope levels. It is used as a histological stain for peroxidase activity. Reaction between 3,3'-Diaminobenzidine and peroxidase forms a visible insoluble product. This approach has been used to assay activity of glutamate decarboxylase in rat cerebellar sections and to detect horseradish peroxidase presence on cultured cell surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3,3'-Diaminobenzidine tetrahydrochloride hydrate is an organic compound used as a peroxidase substrate

Application

• 3,3'-Diaminobenzidine tetrahydrochloride hydrate is commonly used for immunoblotting and immunohistochemical staining techniques

• It has also been used for the histological stain to measure the peroxidase activity. The interaction between 3,3'-Diaminobenzidine tetrahydrochloride hydrate and peroxidase leads to the formation of a visible insoluble product. Therefore, it has been applied to measure glutamate decarboxylase activity in rat cerebellar sections and to detect horseradish peroxidase presence on cultured cell surfaces

• It is used as a chromogen for detecting the antigen-antibody complex

• This compound is also used in several laboratory applications, including electron and light microscope, and FISH (fluorescence in situ hybridization)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868272-85-9
Molecular Formula C12H18Cl4N4
Molecular Weight (g/mol) 360.10
MDL Number MFCD08273058
InChI Key KJDSORYAHBAGPP-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se
PubChem CID 23191111
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate
SMILES Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1

Specifications

Color Gray
Quantity 5 g
Solubility Information Soluble at 5%(w/v) in water
Formula Weight 360.11 (Anhydrous)
Physical Form Powder
Chemical Name or Material 3,3'-Diaminobenzidine tetrahydrochloride hydrate
compliance-icons
Product Identifier
  • 3,3'-Diaminobenzidine tetrahydrochloride hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3,3-Diaminobenzidinetetrahydrochloridehydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'