missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Difluoroazetidine hydrochloride, 95%, Thermo Scientific Chemicals
177.00 € - 500.00 €
Chemical Identifiers
CAS | 288315-03-7 |
---|---|
Molecular Formula | C3H6ClF2N |
MDL Number | MFCD05663714 |
InChI Key | CDBAEFXTCRKJPZ-UHFFFAOYSA-N |
Synonym | 3,3-difluoroazetidine hydrochloride, 3,3-difluoroazetidine hcl, azetidine, 3,3-difluoro-, hydrochloride, azetidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoroazetidine hydrochloride 1:1, 3,3-difluoroazetidinehydrochloride, acmc-1clfn, 3,3-difluoroazetidine-hcl, difluoroazetidine hydrochloride, 3,3-difluoroazetidin hydrochloride |
PubChem CID | 2758247 |
IUPAC Name | 3,3-difluoroazetidine;hydrochloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389959
|
Thermo Scientific Alfa Aesar
H25751.MD |
250 mg |
177.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399959
|
Thermo Scientific Alfa Aesar
H25751.03 |
1 g |
500.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
288315-03-7 | |
MFCD05663714 | |
3,3-difluoroazetidine hydrochloride, 3,3-difluoroazetidine hcl, azetidine, 3,3-difluoro-, hydrochloride, azetidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoroazetidine hydrochloride 1:1, 3,3-difluoroazetidinehydrochloride, acmc-1clfn, 3,3-difluoroazetidine-hcl, difluoroazetidine hydrochloride, 3,3-difluoroazetidin hydrochloride | |
3,3-difluoroazetidine;hydrochloride |
C3H6ClF2N | |
CDBAEFXTCRKJPZ-UHFFFAOYSA-N | |
2758247 |
Specifications
288315-03-7 | |
C3H6ClF2N | |
Hygroscopic | |
CDBAEFXTCRKJPZ-UHFFFAOYSA-N | |
3,3-difluoroazetidine;hydrochloride | |
2758247 | |
95% |
138°C to 140°C | |
MFCD05663714 | |
3,3-difluoroazetidine hydrochloride, 3,3-difluoroazetidine hcl, azetidine, 3,3-difluoro-, hydrochloride, azetidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoroazetidine hydrochloride 1:1, 3,3-difluoroazetidinehydrochloride, acmc-1clfn, 3,3-difluoroazetidine-hcl, difluoroazetidine hydrochloride, 3,3-difluoroazetidin hydrochloride | |
C1C(CN1)(F)F.Cl | |
129.535 | |
129.54 | |
3,3-Difluoroazetidine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only