missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Difluorobiphenyl, 97%, Thermo Scientific Chemicals

97.00 € - 427.00 €

Chemical Identifiers

CAS 396-64-5
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.19
MDL Number MFCD00039216
InChI Key GAYJHUJLHJWCTH-UHFFFAOYSA-N
Synonym 3,3'-difluorobiphenyl, 3,3'-difluoro-1,1'-biphenyl, 1-fluoro-3-3-fluorophenyl benzene, 1,1'-biphenyl,3,3'-difluoro, 1,1'-biphenyl, 3,3'-difluoro, 3-fluoro-1-3-fluorophenyl benzene, acmc-1agdg, 3,3'-difluoro biphenyl, gayjhujlhjwcth-uhfffaoysa, 3,3'-difluoro-1,1'-biphenyl #
PubChem CID 123058
IUPAC Name 1-fluoro-3-(3-fluorophenyl)benzene
SMILES FC1=CC(=CC=C1)C1=CC(F)=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329375
View Documents
Thermo Scientific Alfa Aesar
L16740.03
1 g
97.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11339375
View Documents
Thermo Scientific Alfa Aesar
L16740.06
5 g
427.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3,3'-Difluorobiphenyl is used materials in ferroelectric display devices

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Difluorobiphenyl is used materials in ferroelectric display devices

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

396-64-5
190.19
GAYJHUJLHJWCTH-UHFFFAOYSA-N
123058
FC1=CC(=CC=C1)C1=CC(F)=CC=C1
C12H8F2
MFCD00039216
3,3'-difluorobiphenyl, 3,3'-difluoro-1,1'-biphenyl, 1-fluoro-3-3-fluorophenyl benzene, 1,1'-biphenyl,3,3'-difluoro, 1,1'-biphenyl, 3,3'-difluoro, 3-fluoro-1-3-fluorophenyl benzene, acmc-1agdg, 3,3'-difluoro biphenyl, gayjhujlhjwcth-uhfffaoysa, 3,3'-difluoro-1,1'-biphenyl #
1-fluoro-3-(3-fluorophenyl)benzene

Specifications

396-64-5
1.21
C12H8F2
MFCD00039216
2046445
Difficult to mix.
FC1=CC(=CC=C1)C1=CC(F)=CC=C1
190.19
190.19
3,3'-Difluorobiphenyl
7°C to 8°C
130°C (14 mmHg)
1.568
UN3151
3,3'-difluorobiphenyl, 3,3'-difluoro-1,1'-biphenyl, 1-fluoro-3-3-fluorophenyl benzene, 1,1'-biphenyl,3,3'-difluoro, 1,1'-biphenyl, 3,3'-difluoro, 3-fluoro-1-3-fluorophenyl benzene, acmc-1agdg, 3,3'-difluoro biphenyl, gayjhujlhjwcth-uhfffaoysa, 3,3'-difluoro-1,1'-biphenyl #
GAYJHUJLHJWCTH-UHFFFAOYSA-N
1-fluoro-3-(3-fluorophenyl)benzene
123058
97%
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, LIQUID

missing translation for 'einecsNumber' : 206-905-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.