missing translation for 'onlineSavingsMsg'
Learn More
3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals Product Code.: 10341274

3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals

Product Code. 10341274
25 mL, Glass bottle
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10341274

Brand: Thermo Scientific Acros 223440250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-92-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.13
MDL Number MFCD00008852
InChI Key PPWNCLVNXGCGAF-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
PubChem CID 13512
IUPAC Name 3,3-dimethylbut-1-yne
SMILES CC(C)(C)C#C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -78.0°C
Color Colorless
Density 0.6600g/mL
Boiling Point 33.0°C to 38.0°C
Flash Point −034°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC≡CH
Refractive Index 1.3730 to 1.3750
Quantity 25 mL
Beilstein 01, 256
Specific Gravity 0.66
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,benzene,toluene
Formula Weight 82.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethyl-1-butyne
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466643 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number223440250,223440050
Lot NumberA0441479 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number223440250,223440050
Lot NumberA0457174 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number223440250,223440050
Lot NumberA0399249 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number223440250,223440050
Lot NumberA0409989 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number223440250,223440050
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.