missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals

Product Code. 11974821
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-97-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00008846
InChI Key PJGSXYOJTGTZAV-UHFFFAOYSA-N
Synonym pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone
PubChem CID 6416
IUPAC Name 3,3-dimethylbutan-2-one
SMILES CC(=O)C(C)(C)C

Specifications

Melting Point -52.5°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 106.0°C
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3CCOCH3
Refractive Index 1.3950 to 1.3980
Quantity 2.5 L
UN Number 1993
Beilstein 01, 694
Merck Index 15, 7551
Specific Gravity 0.8
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 100.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethyl-2-butanone, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethyl-2-butanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'