missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dithiodipropionic acid, 98%

Product Code. 10716191 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 10716191

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1119-62-6 | C6H10O4S2 | 210.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-62-6
Molecular Formula C6H10O4S2
Molecular Weight (g/mol) 210.26
MDL Number MFCD00002780
InChI Key YCLSOMLVSHPPFV-UHFFFAOYSA-N
Synonym 3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid
PubChem CID 95116
IUPAC Name 3-(2-carboxyethyldisulfanyl)propanoic acid
SMILES C(CSSCCC(=O)O)C(=O)O

Specifications

CAS Min % 98
Melting Point 155°C to 158°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula S2(CH2CH2CO2H)2
Quantity 50 g
Beilstein 03, IV, 736
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,benzene
Formula Weight 210.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 3'-Dithiodipropionic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,3'-Dithiodipropionic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.