missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-(3-Hydroxypropyl)benzeneboronic acid, 96%
Click to view available options
Quantity:
250 mg
Specifications
Specifications
CAS | 736989-98-3 |
Molecular Formula | C9H13BO3 |
MDL Number | MFCD04115650 |
Quantity | 250 mg |
Synonym | 3-3-hydroxypropyl phenylboronic acid, 3-3-hydroxypropyl phenyl boronic acid, 3-3-boronophenyl propan-1-ol, boronic acid, 3-3-hydroxypropyl phenyl, 3-3-hydroxypropyl benzeneboronic acid, 3-3-hydroxyprop-1-yl benzeneboronic acid, acmc-1bkf7, 3-hydroxypropyl phenylboronic acid, 3-3-hydroxypropyl phenyl boronicacid |
InChI Key | OTCBPHAZRFXGKH-UHFFFAOYSA-N |
SMILES | OCCCC1=CC=CC(=C1)B(O)O |
IUPAC Name | [3-(3-hydroxypropyl)phenyl]boronic acid |
Molecular Weight (g/mol) | 180.01 |
PubChem CID | 3672173 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction