missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Nitrophenyl)-1H-pyrazole, 97%, Thermo Scientific™

57.00 € - 158.79 €

Chemical Identifiers

CAS 59843-77-5
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
MDL Number MFCD00106146
InChI Key RUWXJVIBNZSEQD-UHFFFAOYSA-N
Synonym 3-3-nitrophenyl-1h-pyrazole, 3-3-nitrophenyl pyrazole, 3-3-nitro-phenyl-1h-pyrazole, 5-3-nitrophenyl-1h-pyrazole, 5-3-nitrophenyl pyrazole, 5-3-nitro-phenyl-1h-pyrazole, 3-3-nitrophenyl-2h-pyrazole, 3-nitro phenylpyrazole, pubchem22739
PubChem CID 2737070
IUPAC Name 5-(3-nitrophenyl)-1H-pyrazole
SMILES [O-][N+](=O)C1=CC=CC(=C1)C1=CC=NN1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465227
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32498.ME
500 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15455227
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32498.04
2 g
158.79 €
2g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

59843-77-5
189.17
RUWXJVIBNZSEQD-UHFFFAOYSA-N
2737070
[O-][N+](=O)C1=CC=CC(=C1)C1=CC=NN1
C9H7N3O2
MFCD00106146
3-3-nitrophenyl-1h-pyrazole, 3-3-nitrophenyl pyrazole, 3-3-nitro-phenyl-1h-pyrazole, 5-3-nitrophenyl-1h-pyrazole, 5-3-nitrophenyl pyrazole, 5-3-nitro-phenyl-1h-pyrazole, 3-3-nitrophenyl-2h-pyrazole, 3-nitro phenylpyrazole, pubchem22739
5-(3-nitrophenyl)-1H-pyrazole

Specifications

59843-77-5
C9H7N3O2
3-3-nitrophenyl-1h-pyrazole, 3-3-nitrophenyl pyrazole, 3-3-nitro-phenyl-1h-pyrazole, 5-3-nitrophenyl-1h-pyrazole, 5-3-nitrophenyl pyrazole, 5-3-nitro-phenyl-1h-pyrazole, 3-3-nitrophenyl-2h-pyrazole, 3-nitro phenylpyrazole, pubchem22739
[O-][N+](=O)C1=CC=CC(=C1)C1=CC=NN1
189.17
189.18
3-(3-Nitrophenyl)-1H-pyrazole
126°C to 128°C
MFCD00106146
RUWXJVIBNZSEQD-UHFFFAOYSA-N
5-(3-nitrophenyl)-1H-pyrazole
2737070
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications