missing translation for 'onlineSavingsMsg'
Learn More

3,3-Pentamethylene glutarimide, ≥98%, Thermo Scientific™

Product Code. 15442309
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1130-32-1
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD00023872
InChI Key FNIPRNMPSXNBDI-UHFFFAOYSA-N
Synonym 3,3-pentamethylene glutarimide, 3-azaspiro 5.5 undecane-2,4-dione, 1,1-cyclohexanediacetimide, 3,3-pentamethyleneglutarimide, 3-azaspiro 5,5 undecan-2,4-dione, 9-azaspiro 5.5 undecane-8,10-dione, beta,beta-pentamethylen-glutarsaeureimid german, 5-21-10-00098 beilstein handbook reference, beta,beta-pentamethylen-glutarsaeureimid
PubChem CID 14324
IUPAC Name 3-azaspiro[5.5]undecane-2,4-dione
SMILES C1CCC2(CC1)CC(=O)NC(=O)C2

Specifications

Melting Point 168°C to 170°C
Quantity 1 g
Beilstein 383782
Formula Weight 181.23
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 3,3-Pentamethylene glutarimide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'