missing translation for 'onlineSavingsMsg'
Learn More
3-(4,5-Diphenyl-1,3-oxazol-2-yl)propanoic acid, 97%, Thermo Scientific™ Product Code.: 10703377

3-(4,5-Diphenyl-1,3-oxazol-2-yl)propanoic acid, 97%, Thermo Scientific™

Product Code. 10703377
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10703377

Brand: Thermo Scientific Maybridge SS00042DE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 21256-18-8
Molecular Formula C18H15NO3
Molecular Weight (g/mol) 293.32
MDL Number MFCD00215977
InChI Key OFPXSFXSNFPTHF-UHFFFAOYSA-N
Synonym oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir
PubChem CID 4614
ChEBI CHEBI:7822
IUPAC Name 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
SMILES OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Merck Index 15, 7023
Formula Weight 293.32
Percent Purity 97%
Chemical Name or Material 3-(4, 5-Diphenyl-1, 3-oxazol-2-yl)propanoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(4,5-Diphenyl-1,3-oxazol-2-yl)propanoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.