missing translation for 'onlineSavingsMsg'
Learn More

3',4',7,8-Tetramethoxyflavone, 97%, Thermo Scientific Chemicals

Product Code. 11303277
Click to view available options
Quantity:
50 mg
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65548-55-2
Molecular Formula C19H18O6
Molecular Weight (g/mol) 342.347
MDL Number MFCD00143001
InChI Key ZRLWYUNKZNRQLO-UHFFFAOYSA-N
Synonym 3',4',7,8-tetramethoxyflavone, 2-3,4-dimethoxyphenyl-7,8-dimethoxy-4h-chromen-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxychromen-4-one, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxy, 2-3,4-dimethoxy-phenyl-7,8-dimethoxy-chromen-4-one, acmc-1b8pz, 7,8,3',4'-tetramethoxyflavone, 7,8-dimethoxy-2-3,4-dimethoxyphenyl-4h-chromen-4-one
PubChem CID 4033898
IUPAC Name 2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)OC)OC)OC

Specifications

Melting Point 206°C to 208°C
Quantity 50 mg
Beilstein 330821
Formula Weight 342.35
Percent Purity 97%
Chemical Name or Material 3',4',7,8-Tetramethoxyflavone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'