missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Benzyloxy)phenyl]propionic acid, 96%, Thermo Scientific Chemicals

Código de producto. 15416627
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 50463-48-4
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD01570264
InChI Key QTSAUVQZNADEKS-UHFFFAOYSA-N
Synonym 3-4-benzyloxyphenyl propionic acid, 3-4-benzyloxy phenyl propanoic acid, 3-4-benzyloxy phenyl propionic acid, benzyloxyphenylpropanoic acid, 3-4-benzyloxyphenyl propanoic acid, 3-4-benzyloxy-phenyl-propionic acid, benzenepropanoic acid, 4-phenylmethoxy, maybridge3_003480, 4-benzyloxy benzenepropanoic acid
PubChem CID 561372
IUPAC Name 3-(4-phenylmethoxyphenyl)propanoic acid
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CCC(=O)O

Especificaciones

Melting Point 116°C to 118°C
Quantity 5 g
Formula Weight 256.3
Percent Purity 96%
Chemical Name or Material 3-[4-(Benzyloxy)phenyl]propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'