missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenoxy)-N,N-dimethylpropylamine, 97%, Thermo Scientific™

Product Code. 10205794
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 76579-64-1
Molecular Formula C11H16BrNO
Molecular Weight (g/mol) 258.159
MDL Number MFCD03718813
InChI Key CAPNMXRLKVMOOB-UHFFFAOYSA-N
Synonym 3-4-bromophenoxy propyl dimethylamine, 3-4-bromophenoxy-n,n-dimethylpropylamine, 3-4-bromophenoxy-n,n-dimethylpropan-1-amine, 1-propanamine,3-4-bromophenoxy-n,n-dimethyl, 4-n,n-dimethylaminopropyloxy phenyl bromide, n,n-dimethyl-3-4-bromophenoxy-1-propanamine
PubChem CID 2183674
IUPAC Name 3-(4-bromophenoxy)-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCOC1=CC=C(C=C1)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 258.16
Percent Purity 97%
Chemical Name or Material 3-(4-Bromophenoxy)-N, N-dimethylpropylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'