missing translation for 'onlineSavingsMsg'
Learn More

3-[4-Chloro-2-(trifluoromethyl)phenyl]propionic acid, 95%, Thermo Scientific™

Product Code. 15418095
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-46-7
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.617
MDL Number MFCD09025363
InChI Key OWXLPJQRNXEXHZ-UHFFFAOYSA-N
Synonym 3-4-chloro-2-trifluoromethyl phenyl propionic acid, 3-4-chloro-2-trifluoromethyl phenyl propanoic acid
PubChem CID 46737495
IUPAC Name 3-[4-chloro-2-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)CCC(=O)O

Specifications

Melting Point 87°C to 90°C
Quantity 1 g
Formula Weight 252.62
Percent Purity 95%
Chemical Name or Material 3-[4-Chloro-2-(trifluoromethyl)phenyl]propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'