missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chloro-3-fluorophenyl)propionic acid, 96%, Thermo Scientific Chemicals

Product Code. 15447137
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881189-65-7
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.609
MDL Number MFCD04116058
InChI Key GDXMVKNNJGWZFN-UHFFFAOYSA-N
Synonym 3-4-chloro-3-fluorophenyl propionic acid, 3-4-chloro-3-fluorophenyl propanoic acid, benzenepropanoic acid, 4-chloro-3-fluoro, 3-4-chloro-3-fluoro-phenyl-propionic acid, 4-chloro-3-fluorobenzenepropanoic acid, 3-4-chloro-3-fluorophenyl propionicacid
PubChem CID 4680108
IUPAC Name 3-(4-chloro-3-fluorophenyl)propanoic acid
SMILES C1=CC(=C(C=C1CCC(=O)O)F)Cl

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
Formula Weight 202.61
Percent Purity 96%
Chemical Name or Material 3-(4-Chloro-3-fluorophenyl)propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'