missing translation for 'onlineSavingsMsg'
Learn More
3,4-Diaminophenylboronic acid pinacol ester, 97% Product Code.: 11996071

3,4-Diaminophenylboronic acid pinacol ester, 97%

Product Code. 11996071
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 11996071

Brand: Thermo Scientific Acros 444760010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 851883-08-4 | C12H19BN2O2 | 234.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 851883-08-4
Melting Point 95°C to 97°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C12H19BN2O2
MDL Number MFCD09027073
Quantity 1 g
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,2-diamine, 3,4-diaminophenylboronic acid, pinacol ester, 3,4-diaminophenylboronic acid pinacol ester, pubchem22338, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzene-1,2-diamine, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-benzenediamine, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,2-diamine
InChI Key SVUDHDDKOKWBNK-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC(N)=C(N)C=C1
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine
Molecular Weight (g/mol) 234.11
PubChem CID 17750242
Formula Weight 234.11
Percent Purity 97%
Chemical Name or Material 3, 4-Diaminophenylboronic acid pinacol ester
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345382 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number444760010
compliance-icons
Product Identifier
  • 3,4-Diaminophenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Diaminophenylboronic acid pinacol ester, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.