Learn More
3,4-Diaminopyridine, 98%, Thermo Scientific Chemicals
82.41 € - 295.47 €
Chemical Identifiers
CAS | 54-96-6 |
---|---|
Molecular Formula | C5H7N3 |
Molecular Weight (g/mol) | 109.13 |
MDL Number | MFCD00006401 |
InChI Key | OYTKINVCDFNREN-UHFFFAOYSA-N |
Synonym | 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino |
PubChem CID | 5918 |
IUPAC Name | pyridine-3,4-diamine |
SMILES | C1=CN=CC(=C1N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10346320
|
Thermo Scientific
112390050 |
5 g | Glass bottle |
82.41 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10542364
|
Thermo Scientific Acros
112390250 |
25 g | Glass bottle |
295.47 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54-96-6 | |
109.13 | |
OYTKINVCDFNREN-UHFFFAOYSA-N | |
5918 | |
C1=CN=CC(=C1N)N |
C5H7N3 | |
MFCD00006401 | |
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino | |
pyridine-3,4-diamine |
Specifications
54-96-6 | |
100.0 | |
Brown to Brown-Gray | |
Authentic | |
C5H7N3 | |
MFCD00006401 | |
15, 398 | |
Solubility in water: 24g/L (20°C) | |
C1=CN=CC(=C1N)N | |
109.13 | |
109.13 | |
Crystalline Powder |
97.5 | |
216°C to 221°C | |
240°C | |
97.5% min. (GC) | |
(C5H3N)(NH2)2 | |
22, II, 395 | |
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino | |
OYTKINVCDFNREN-UHFFFAOYSA-N | |
pyridine-3,4-diamine | |
5918 | |
98% | |
3, 4-Diaminopyridine |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-220-9
Recommended Storage : Product darkens on storage
- 3,4-Diaminopyridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H330-Fatal if inhaled.
- H311-Toxic in contact with skin.
- H335-May cause respiratory irritation.
- H300-Fatal if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.