missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dichlorophenyl isocyanate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C 'labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C ′″labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, amines, alcohols.
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C ′″labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, amines, alcohols.
Specifications
Specifications
Melting Point | 40°C to 44°C |
Boiling Point | 118°C to 120°C (18 mmHg) |
Flash Point | 123°C (253°F) |
Odor | Irritating |
Quantity | 25 g |
UN Number | UN2250 |
Beilstein | 608325 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 188.01 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'