missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl, 97%, Thermo Scientific™

64.25 € - 251.00 €

Chemical Identifiers

CAS 134412-18-3
Molecular Formula C20H22F2
Molecular Weight (g/mol) 300.39
MDL Number MFCD12828121
InChI Key TYZNCUASDJPXMP-UHFFFAOYSA-N
Synonym 3,4-difluoro-4'-4-ethylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-ethylcyclohexyl biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro-1,1'-biphenyl, 3,4-difluoro-4/'-4-ethylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-ethylcyclohexyl benzene, 4-4-4-ethylcyclohexyl phenyl-1,2-difluorobenzene, 1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro, 4'-4alpha-ethylcyclohexane-1beta-yl-3,4-difluorobiphenyl, 4-4-ethylcyclohexyl-3',4'-difluoro-1,1'-biphenyl #
PubChem CID 612308
SMILES CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431478
View Documents
Thermo Scientific Alfa Aesar
H55275.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15441478
View Documents
Thermo Scientific Alfa Aesar
H55275.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

134412-18-3
300.39
TYZNCUASDJPXMP-UHFFFAOYSA-N
612308
C20H22F2
MFCD12828121
3,4-difluoro-4'-4-ethylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-ethylcyclohexyl biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro-1,1'-biphenyl, 3,4-difluoro-4/'-4-ethylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-ethylcyclohexyl benzene, 4-4-4-ethylcyclohexyl phenyl-1,2-difluorobenzene, 1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro, 4'-4alpha-ethylcyclohexane-1beta-yl-3,4-difluorobiphenyl, 4-4-ethylcyclohexyl-3',4'-difluoro-1,1'-biphenyl #
CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

134412-18-3
C20H22F2
3,4-difluoro-4'-4-ethylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-ethylcyclohexyl biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro-1,1'-biphenyl, 3,4-difluoro-4/'-4-ethylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-ethylcyclohexyl benzene, 4-4-4-ethylcyclohexyl phenyl-1,2-difluorobenzene, 1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro, 4'-4alpha-ethylcyclohexane-1beta-yl-3,4-difluorobiphenyl, 4-4-ethylcyclohexyl-3',4'-difluoro-1,1'-biphenyl #
CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1
300.39
300.39
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl
68°C to 69°C
MFCD12828121
TYZNCUASDJPXMP-UHFFFAOYSA-N
4'-(4-ethylcyclohexyl)-3,4-difluoro-1,1'-biphenyl
612308
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.