missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenol, 98%, Thermo Scientific Chemicals

32.50 € - 327.00 €

Chemical Identifiers

CAS 2713-33-9
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00010315
InChI Key BNPWVUJOPCGHIK-UHFFFAOYSA-N
Synonym phenol, 3,4-difluoro, 3,4-difluoro phenol, 3 4-difluorophenol, 3,4-difluoro-phenol, pubchem2315, acmc-1cmaj, 3,4-difluorophenol, ksc202q5j, bnpwvujopcghik-uhfffaoysa
PubChem CID 75927
IUPAC Name 3,4-difluorophenol
SMILES OC1=CC=C(F)C(F)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11404324
View Documents
Thermo Scientific Alfa Aesar
A12596.06
5 g
32.50 €
5g
Please to purchase this item. Need a web account? Register with us today!
11414324
View Documents
Thermo Scientific Alfa Aesar
A12596.14
25 g
106.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
11424324
View Documents
Thermo Scientific Alfa Aesar
A12596.22
100 g
327.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2713-33-9
130.09
BNPWVUJOPCGHIK-UHFFFAOYSA-N
75927
OC1=CC=C(F)C(F)=C1
C6H4F2O
MFCD00010315
phenol, 3,4-difluoro, 3,4-difluoro phenol, 3 4-difluorophenol, 3,4-difluoro-phenol, pubchem2315, acmc-1cmaj, 3,4-difluorophenol, ksc202q5j, bnpwvujopcghik-uhfffaoysa
3,4-difluorophenol

Specifications

2713-33-9
84°C to 86°C (20 mmHg)
Phenol-like
MFCD00010315
2081085
BNPWVUJOPCGHIK-UHFFFAOYSA-N
3,4-difluorophenol
75927
98%
34°C to 38°C
57°C (134°F)
C6H4F2O
UN2921
phenol, 3,4-difluoro, 3,4-difluoro phenol, 3 4-difluorophenol, 3,4-difluoro-phenol, pubchem2315, acmc-1cmaj, 3,4-difluorophenol, ksc202q5j, bnpwvujopcghik-uhfffaoysa
OC1=CC=C(F)C(F)=C1
130.09
130.09
3,4-Difluorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H228-H302+H312+H332-H314

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III

missing translation for 'einecsNumber' : 424-300-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.