missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid, ≥97%, Thermo Scientific™ Product Code.: 10669364

3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10669364
Amber Glass Bottle; 250mg
Click to view available options
Amber Glass Bottle; 250mg
This item is not returnable. View return policy

Product Code. 10669364

Brand: Thermo Scientific Maybridge CC29701CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 90563-93-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H9NO3
MDL Number MFCD05865108
Quantity 250mg
Synonym 2h-1,4-benzoxazine-2-carboxylicacid, 3,4-dihydro, 2h-1,4-benzoxazine-2-carboxylicacid,3,4-dihydro, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, benzomorpholine-2-carboxylic acid, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carboxylic acid, +/--3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid
InChI Key VNTNLTOJGXENIZ-UHFFFAOYSA-N
SMILES C1C(OC2=CC=CC=C2N1)C(=O)O
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Molecular Weight (g/mol) 179.175
PubChem CID 3826952
Formula Weight 179.17
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.