missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzaldoxime, 98%, Thermo Scientific™

Product Code. 11314755
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3343-59-7
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD01070622
InChI Key XQTLVFXPCMGDOB-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzaldoxime, 3,4-dihydroxybenzaldehyde oxime, acmc-20anf4, 4-hydroxyimino methyl benzene-1,2-diol, 3,4-dihydroxybenzaldehydeoxime, nufmsecagoxhav-uhfffaoysa-n, benzaldehyde,3,4-dihydroxy-, oxime, 4-1e-hydroxyimino methyl benzene-1,2-diol, 2-hydroxy-4-hydroxyamino methylidene cyclohexa-2,5-dien-1-one
PubChem CID 6798780
IUPAC Name 2-hydroxy-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one
SMILES C1=CC(=O)C(=CC1=CNO)O

Specifications

Melting Point 155°C to 157°C (decomposition)
Quantity 5 g
Beilstein 3126402
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3,4-Dihydroxybenzaldoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'