Learn More
3',4'-Dihydroxypropiophenone, 98%, Thermo Scientific Chemicals
39.70 € - 144.00 €
Chemical Identifiers
CAS | 7451-98-1 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.176 |
MDL Number | MFCD00016440 |
InChI Key | HNWIHBDMOYWCGX-UHFFFAOYSA-N |
Synonym | 3',4'-dihydroxypropiophenone, 3,4-dihydroxypropiophenone, propiophenone, 3',4'-dihydroxy, 1-3,4-dihydroxyphenyl propan-1-one, 1-propanone, 1-3,4-dihydroxyphenyl, 1-3,4-dihydroxyphenyl-1-propanone, 1-propanone,1-3,4-dihydroxyphenyl, 4-propionylbrenzcatechin, propiophenone,4'-dihydroxy, 3', 4'-dihydroxypropiophenone |
PubChem CID | 96267 |
IUPAC Name | 1-(3,4-dihydroxyphenyl)propan-1-one |
SMILES | CCC(=O)C1=CC(=C(C=C1)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11415503
|
Thermo Scientific Alfa Aesar
A13444.03 |
1 g |
39.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425503
|
Thermo Scientific Alfa Aesar
A13444.06 |
5 g |
144.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
7451-98-1 | |
166.176 | |
HNWIHBDMOYWCGX-UHFFFAOYSA-N | |
96267 | |
CCC(=O)C1=CC(=C(C=C1)O)O |
C9H10O3 | |
MFCD00016440 | |
3',4'-dihydroxypropiophenone, 3,4-dihydroxypropiophenone, propiophenone, 3',4'-dihydroxy, 1-3,4-dihydroxyphenyl propan-1-one, 1-propanone, 1-3,4-dihydroxyphenyl, 1-3,4-dihydroxyphenyl-1-propanone, 1-propanone,1-3,4-dihydroxyphenyl, 4-propionylbrenzcatechin, propiophenone,4'-dihydroxy, 3', 4'-dihydroxypropiophenone | |
1-(3,4-dihydroxyphenyl)propan-1-one |
Specifications
7451-98-1 | |
C9H10O3 | |
3',4'-dihydroxypropiophenone, 3,4-dihydroxypropiophenone, propiophenone, 3',4'-dihydroxy, 1-3,4-dihydroxyphenyl propan-1-one, 1-propanone, 1-3,4-dihydroxyphenyl, 1-3,4-dihydroxyphenyl-1-propanone, 1-propanone,1-3,4-dihydroxyphenyl, 4-propionylbrenzcatechin, propiophenone,4'-dihydroxy, 3', 4'-dihydroxypropiophenone | |
CCC(=O)C1=CC(=C(C=C1)O)O | |
166.176 | |
166.18 | |
3',4'-Dihydroxypropiophenone |
145°C to 146°C | |
MFCD00016440 | |
HNWIHBDMOYWCGX-UHFFFAOYSA-N | |
1-(3,4-dihydroxyphenyl)propan-1-one | |
96267 | |
98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only