missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

14.37 € - 277.38 €

Chemical Identifiers

CAS 1131-62-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00008737
InChI Key IQZLUWLMQNGTIW-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
PubChem CID 14328
ChEBI CHEBI:86576
IUPAC Name 1-(3,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)OC
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10612413
View Documents
Thermo Scientific Acros
168850050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10406432
View Documents
Thermo Scientific Acros
168850250
25 g Glass bottle
33.57 €
25g
Please to purchase this item. Need a web account? Register with us today!
10275930
View Documents
Thermo Scientific Acros
168851000
100 g Glass bottle
80.60 €
100g
Please to purchase this item. Need a web account? Register with us today!
10076093
View Documents
Thermo Scientific Acros
168855000
500 g Glass bottle
277.38 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1131-62-0
180.2
IQZLUWLMQNGTIW-UHFFFAOYSA-N
14328
1-(3,4-dimethoxyphenyl)ethanone
C10H12O3
MFCD00008737
3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
CHEBI:86576
CC(=O)C1=CC(=C(C=C1)OC)OC

Specifications

1131-62-0
100.0
Yellow to Beige
>112°C
97.5% min. (GC)
(CH3O)2C6H3COCH3
08, II, 298
Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol
CC(=O)C1=CC(=C(C=C1)OC)OC
180.2
CHEBI:86576
98%
3', 4'-Dimethoxyacetophenone
97.5
48.0°C to 52.0°C
286.0°C to 288.0°C
Authentic
C10H12O3
MFCD00008737
3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
IQZLUWLMQNGTIW-UHFFFAOYSA-N
1-(3,4-dimethoxyphenyl)ethanone
14328
180.2
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-468-

Product Identifier
  • 3',4'-Dimethoxyacetophenone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.