Learn More
3,4-Dimethylphenol, 99%, Thermo Scientific Chemicals
32.36 € - 318.59 €
Chemical Identifiers
CAS | 95-65-8 |
---|---|
Molecular Formula | C8H10O |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00002304 |
InChI Key | YCOXTKKNXUZSKD-UHFFFAOYSA-N |
Synonym | 3,4-xylenol, phenol, 3,4-dimethyl, 4,5-dimethylphenol, 3,4-dimethyl phenol, 1,3,4-xylenol, 4-hydroxy-1,2-dimethylbenzene, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl-phenol, 3,4-dmp, ccris 723 |
PubChem CID | 7249 |
ChEBI | CHEBI:39839 |
IUPAC Name | 3,4-dimethylphenol |
SMILES | CC1=CC=C(O)C=C1C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10386740
|
Thermo Scientific Acros
116581000 |
100 g | Glass bottle |
32.36 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10530741
|
Thermo Scientific Acros
116585000 |
500 g | Glass bottle |
97.79 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10793455
|
Thermo Scientific Acros
116580025 |
2.5 kg | Glass bottle |
318.59 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-65-8 | |
122.17 | |
YCOXTKKNXUZSKD-UHFFFAOYSA-N | |
7249 | |
3,4-dimethylphenol |
C8H10O | |
MFCD00002304 | |
3,4-xylenol, phenol, 3,4-dimethyl, 4,5-dimethylphenol, 3,4-dimethyl phenol, 1,3,4-xylenol, 4-hydroxy-1,2-dimethylbenzene, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl-phenol, 3,4-dmp, ccris 723 | |
CHEBI:39839 | |
CC1=CC=C(O)C=C1C |
Specifications
95-65-8 | |
100.0 | |
Cream to White | |
227°C | |
Authentic | |
C8H10O | |
MFCD00002304 | |
15, 10279 | |
3,4-xylenol, phenol, 3,4-dimethyl, 4,5-dimethylphenol, 3,4-dimethyl phenol, 1,3,4-xylenol, 4-hydroxy-1,2-dimethylbenzene, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl-phenol, 3,4-dmp, ccris 723 | |
YCOXTKKNXUZSKD-UHFFFAOYSA-N | |
3,4-dimethylphenol | |
7249 | |
122.17 | |
Crystalline Powder |
98.5 | |
63°C to 67°C | |
0.9830g/mL | |
80°C | |
98.5% min. (GC) | |
(CH3)2C6H3OH | |
06, 480 | |
0.983 | |
Solubility in water: slightly soluble. Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene | |
CC1=CC=C(O)C=C1C | |
122.17 | |
CHEBI:39839 | |
99% | |
3, 4-Dimethylphenol, 99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-439-5
missing translation for 'rtecsNumber' : ZE6300000
missing translation for 'tsca' : TSCA
- 3,4-Dimethylphenol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 3,4-Dimethylphenol
RUO – Research Use Only