Learn More
3-(4-Hydroxyphenyl)propionic acid, 99%, Thermo Scientific Chemicals
34.07 € - 409.04 €
Chemical Identifiers
CAS | 501-97-3 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.176 |
MDL Number | MFCD00002778 |
InChI Key | NMHMNPHRMNGLLB-UHFFFAOYSA-N |
Synonym | 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid |
PubChem CID | 10394 |
ChEBI | CHEBI:32980 |
IUPAC Name | 3-(4-hydroxyphenyl)propanoic acid |
SMILES | C1=CC(=CC=C1CCC(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11431958
|
Thermo Scientific Alfa Aesar
A14567.09 |
10 g |
34.07 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441958
|
Thermo Scientific Alfa Aesar
A14567.18 |
50 g |
91.25 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451958
|
Thermo Scientific Alfa Aesar
A14567.30 |
250 g |
409.04 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
501-97-3 | |
166.176 | |
NMHMNPHRMNGLLB-UHFFFAOYSA-N | |
10394 | |
3-(4-hydroxyphenyl)propanoic acid |
C9H10O3 | |
MFCD00002778 | |
3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid | |
CHEBI:32980 | |
C1=CC(=CC=C1CCC(=O)O)O |
Specifications
501-97-3 | |
C9H10O3 | |
2209841 | |
Soluble in water. | |
C1=CC(=CC=C1CCC(=O)O)O | |
166.176 | |
CHEBI:32980 | |
99% |
126°C to 131°C | |
MFCD00002778 | |
3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid | |
NMHMNPHRMNGLLB-UHFFFAOYSA-N | |
3-(4-hydroxyphenyl)propanoic acid | |
10394 | |
166.18 | |
3-(4-Hydroxyphenyl)propionic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 207-931-3
missing translation for 'rtecsNumber' : MW5342000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only