Learn More
3-(4-Hydroxyphenyl)propionitrile, 98%, Thermo Scientific Chemicals
44.40 € - 146.00 €
Chemical Identifiers
CAS | 17362-17-3 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.18 |
MDL Number | MFCD00016492 |
InChI Key | KDMJGLYRWRHKJS-UHFFFAOYSA-N |
Synonym | 3-4-hydroxyphenyl propionitrile, 3-4-hydroxyphenyl propanenitrile, benzenepropanenitrile, 4-hydroxy, 3-p-hydroxyphenyl propiononitrile, 3-4-hydroxy-phenyl-propionitrile, .beta.-4-hydroxyphenyl propionitrile, pubchem13636, acmc-1bvt5, 4-hydroxyhydrocinnamonitrile, 4-hydroxyphenylpropionitrile |
PubChem CID | 87079 |
IUPAC Name | 3-(4-hydroxyphenyl)propanenitrile |
SMILES | OC1=CC=C(CCC#N)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322107
|
Thermo Scientific Alfa Aesar
L07934.03 |
1 g |
44.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332107
|
Thermo Scientific Alfa Aesar
L07934.06 |
5 g |
146.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
17362-17-3 | |
147.18 | |
KDMJGLYRWRHKJS-UHFFFAOYSA-N | |
87079 | |
OC1=CC=C(CCC#N)C=C1 |
C9H9NO | |
MFCD00016492 | |
3-4-hydroxyphenyl propionitrile, 3-4-hydroxyphenyl propanenitrile, benzenepropanenitrile, 4-hydroxy, 3-p-hydroxyphenyl propiononitrile, 3-4-hydroxy-phenyl-propionitrile, .beta.-4-hydroxyphenyl propionitrile, pubchem13636, acmc-1bvt5, 4-hydroxyhydrocinnamonitrile, 4-hydroxyphenylpropionitrile | |
3-(4-hydroxyphenyl)propanenitrile |
Specifications
17362-17-3 | |
166°C (8 mmHg) | |
MFCD00016492 | |
1448680 | |
Insoluble in water. | |
OC1=CC=C(CCC#N)C=C1 | |
147.18 | |
147.18 | |
3-(4-Hydroxyphenyl)propionitrile |
54°C to 58°C | |
C9H9NO | |
UN3439 | |
3-4-hydroxyphenyl propionitrile, 3-4-hydroxyphenyl propanenitrile, benzenepropanenitrile, 4-hydroxy, 3-p-hydroxyphenyl propiononitrile, 3-4-hydroxy-phenyl-propionitrile, .beta.-4-hydroxyphenyl propionitrile, pubchem13636, acmc-1bvt5, 4-hydroxyhydrocinnamonitrile, 4-hydroxyphenylpropionitrile | |
KDMJGLYRWRHKJS-UHFFFAOYSA-N | |
3-(4-hydroxyphenyl)propanenitrile | |
87079 | |
98% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 241-393-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only