missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Methylbenzoyl)propionic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field
Solubility
Insoluble in water.
Notes
Incompatible with bases and oxidizing agents. Store in a cool, dry condition in well sealed containers.
3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field
Solubility
Insoluble in water.
Notes
Incompatible with bases and oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Specifications
Melting Point | 124°C to 128°C |
Quantity | 25 g |
Beilstein | 2099653 |
Solubility Information | Insoluble in water. |
Formula Weight | 192.22 |
Percent Purity | 98% |
Chemical Name or Material | 3- (4-Methylbenzoyl)propionic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction