missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)propiophenone hydrochloride, 98+%, Thermo Scientific™

Product Code. 11440098
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1020-16-2
Molecular Formula C13H18ClNO2
Molecular Weight (g/mol) 255.742
MDL Number MFCD00035332
InChI Key BEUAFINDWSQCGS-UHFFFAOYSA-N
Synonym 3-morpholinopropiophenone hydrochloride, 3-4-morpholino propiophenone hydrochloride, 3-4-morpholinyl propiophenone hydrochloride, 3-morpholino-1-phenylpropan-1-one hydrochloride, labotest-bb lt00454997, 3-4-morpholinyl-1-phenyl-1-propanone hydrochloride, 3-morpholin-4-yl-1-phenylpropan-1-one hydrochloride, 3-morpholinopropiophenone hcl
PubChem CID 3083670
IUPAC Name 3-morpholin-4-yl-1-phenylpropan-1-one;hydrochloride
SMILES C1COCCN1CCC(=O)C2=CC=CC=C2.Cl

Specifications

Melting Point 180°C to 182°C
Quantity 100 g
Beilstein 3723608
Sensitivity Hygroscopic
Formula Weight 255.76
Percent Purity ≥98%
Chemical Name or Material 3-(4-Morpholinyl)propiophenone hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.