missing translation for 'onlineSavingsMsg'
Learn More

3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10311143
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 500013-38-7
Molecular Formula C17H24N2O4
Molecular Weight (g/mol) 320.389
MDL Number MFCD04115219
InChI Key CXJBKOKDFABTQO-UHFFFAOYSA-N
Synonym 3-4-tert-butoxycarbonyl piperazin-1-yl methyl benzoic acid, 1-3-carboxyphenylmethyl-4-boc piperazine, 1-boc-4-3-carboxy-benzyl-piperazine, 3-4-n-boc-piperazin-1-yl methylbenzoic acid, 1-tert-butoxycarbonyl-4-3-carboxybenzyl piperazine, tert-butyl 4-3-carboxybenzyl piperazine-1-carboxylate, 3-4-tert-butoxy carbonyl piperazin-1-yl methyl benzoic acid, 3-4-tert-butoxycarbonyl piperazin-1-yl methyl benzoicacid, 1-piperazinecarboxylicacid, 4-3-carboxyphenyl methyl-, 1-1,1-dimethylethyl ester
PubChem CID 2795521
IUPAC Name 3-[[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]methyl]benzoic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 320.39
Percent Purity 97%
Chemical Name or Material 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'