missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Spezifikation
Spezifikation
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 320.39 |
Percent Purity | 97% |
Chemical Name or Material | 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'