missing translation for 'onlineSavingsMsg'
Learn More

3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine, 97%, Thermo Scientific™

Product Code. 10107392
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-86-1
Molecular Formula C10H14BNO2
Molecular Weight (g/mol) 191.04
MDL Number MFCD03425979
InChI Key HDQULWCYQQMIJJ-UHFFFAOYSA-N
Synonym 3-5,5-dimethyl-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid neopentylglycol ester, pyridine-3-boronic acid neopentylglycol ester, pyridine-3-boronic acid, neopentyl ester, pyridine-3-boronic acid neopentyl glycol ester, 2-3-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, 5,5-dimethyl-2-pyridin-3-yl-1,3,2-dioxaborinane, 2-4-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, pyridine-3-boronic acid neopentyl ester, pyridine-3-boronic acid, neopentylglycol ester
PubChem CID 2794717
IUPAC Name 3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
SMILES CC1(C)COB(OC1)C1=CC=NC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 191.04
Percent Purity 97%
Chemical Name or Material 3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.