missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15401699
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177735-55-6
Molecular Formula C10H11BO6
Molecular Weight (g/mol) 238.00
MDL Number MFCD11053854
InChI Key WEJWFDLAZSVCJK-UHFFFAOYSA-N
Synonym 3,5-bis methoxycarbonyl phenylboronic acid, 3,5-bis methoxycarbonyl phenyl boronic acid, 3,5-bis methoxycarbonyl benzeneboronic acid, 1,3-benzenedicarboxylic acid, 5-borono-, 1,3-dimethyl ester, 3,5-di methoxycarbonyl phenylboronic acid, acmc-1c6yh, 3,5-bis methoxycarbonyl phenyl-dihydroxyborane
PubChem CID 19363104
IUPAC Name [3,5-bis(methoxycarbonyl)phenyl]boronic acid
SMILES COC(=O)C1=CC(=CC(=C1)B(O)O)C(=O)OC

Specifications

Quantity 1 g
Formula Weight 238
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,5-Bis(methoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.