missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15448946
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177735-55-6
Molecular Formula C10H11BO6
Molecular Weight (g/mol) 238.00
MDL Number MFCD11053854
InChI Key WEJWFDLAZSVCJK-UHFFFAOYSA-N
Synonym 3,5-bis methoxycarbonyl phenylboronic acid, 3,5-bis methoxycarbonyl phenyl boronic acid, 3,5-bis methoxycarbonyl benzeneboronic acid, 1,3-benzenedicarboxylic acid, 5-borono-, 1,3-dimethyl ester, 3,5-di methoxycarbonyl phenylboronic acid, acmc-1c6yh, 3,5-bis methoxycarbonyl phenyl-dihydroxyborane
PubChem CID 19363104
IUPAC Name [3,5-bis(methoxycarbonyl)phenyl]boronic acid
SMILES COC(=O)C1=CC(=CC(=C1)B(O)O)C(=O)OC

Specifications

Quantity 250 mg
Formula Weight 238
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,5-Bis(methoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'