missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. 11308756
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20857-47-0
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD00128904
InChI Key RQEOYYWUVYZZLL-UHFFFAOYSA-N
PubChem CID 2778311
IUPAC Name 3,5-bis(trifluoromethyl)pyridine
SMILES C1=C(C=NC=C1C(F)(F)F)C(F)(F)F

Specifications

Melting Point 35°C to 38°C
Boiling Point 117°C to 118°C
Flash Point >100°C (212°F)
Quantity 1 g
Beilstein 1684576
Synonym 3,5-bis trifluoromethyl pyridine, pyridine, 3,5-bis trifluoromethyl, pubchem7698, acmc-1bncl, 3,5-bis-trifluoromethylpyridine, pyridine,3,5-bis trifluoromethyl
Formula Weight 215.1
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'