missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Bromo-pyridin-3-yl)-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester, ≥97%, Thermo Scientific™

Product Code. 10299872
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-34-7
Molecular Formula C10H8BrN3O3
Molecular Weight (g/mol) 298.096
MDL Number MFCD06797495
InChI Key LSHJXSGSPMZMFQ-UHFFFAOYSA-N
Synonym ethyl 3-5-bromopyridin-3-yl-1,2,4-oxadiazole-5-carboxylate, 3-5-bromo-pyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester, 1,2,4-oxadiazole-5-carboxylicacid, 3-5-bromo-3-pyridinyl-, ethyl ester, 3-5-bromopyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester
PubChem CID 7170018
IUPAC Name ethyl 3-(5-bromopyridin-3-yl)-1,2,4-oxadiazole-5-carboxylate
SMILES CCOC(=O)C1=NC(=NO1)C2=CC(=CN=C2)Br

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 298.1
Percent Purity 95%
Chemical Name or Material 3-(5-Bromo-pyridin-3-yl)-[1, 2, 4]oxadiazole-5-carboxylic acid ethyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(5-Bromo-pyridin-3-yl)-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.