Learn More
3,5-Dibromo-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals
62.60 € - 200.00 €
Chemical Identifiers
CAS | 3337-62-0 |
---|---|
Molecular Formula | C7H4Br2O3 |
Molecular Weight (g/mol) | 295.914 |
MDL Number | MFCD00002548 |
InChI Key | PHWAJJWKNLWZGJ-UHFFFAOYSA-N |
Synonym | bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009 |
PubChem CID | 76857 |
ChEBI | CHEBI:1395 |
IUPAC Name | 3,5-dibromo-4-hydroxybenzoic acid |
SMILES | C1=C(C=C(C(=C1Br)O)Br)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11463927
|
Thermo Scientific Alfa Aesar
A18361.14 |
25 g |
62.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473927
|
Thermo Scientific Alfa Aesar
A18361.22 |
100 g |
200.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3337-62-0 | |
295.914 | |
PHWAJJWKNLWZGJ-UHFFFAOYSA-N | |
76857 | |
3,5-dibromo-4-hydroxybenzoic acid |
C7H4Br2O3 | |
MFCD00002548 | |
bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009 | |
CHEBI:1395 | |
C1=C(C=C(C(=C1Br)O)Br)C(=O)O |
Specifications
3337-62-0 | |
C7H4Br2O3 | |
2416147 | |
PHWAJJWKNLWZGJ-UHFFFAOYSA-N | |
3,5-dibromo-4-hydroxybenzoic acid | |
76857 | |
295.93 | |
3,5-Dibromo-4-hydroxybenzoic acid |
269°C to 273°C | |
MFCD00002548 | |
bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009 | |
C1=C(C=C(C(=C1Br)O)Br)C(=O)O | |
295.914 | |
CHEBI:1395 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 222-075-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 3,5-Dibromo-4-hydroxybenzoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 3,5-Dibromo-4-hydroxybenzoic acid
RUO – Research Use Only