Learn More
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%, Thermo Scientific Chemicals
51.26 € - 909.53 €
Chemical Identifiers
CAS | 54970-72-8 |
---|---|
Molecular Formula | C6H3Cl2NaO4S |
Molecular Weight (g/mol) | 265.036 |
MDL Number | MFCD00009798 |
InChI Key | NMWCVZCSJHJYFW-UHFFFAOYSA-M |
Synonym | sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt |
PubChem CID | 3085077 |
IUPAC Name | sodium;3,5-dichloro-2-hydroxybenzenesulfonate |
SMILES | C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479602
|
Thermo Scientific Alfa Aesar
A10732.09 |
10 g |
51.26 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489602
|
Thermo Scientific Alfa Aesar
A10732.18 |
50 g |
200.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499602
|
Thermo Scientific Alfa Aesar
A10732.30 |
250 g |
909.53 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54970-72-8 | |
265.036 | |
NMWCVZCSJHJYFW-UHFFFAOYSA-M | |
3085077 | |
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+] |
C6H3Cl2NaO4S | |
MFCD00009798 | |
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt | |
sodium;3,5-dichloro-2-hydroxybenzenesulfonate |
Specifications
54970-72-8 | |
C6H3Cl2NaO4S | |
3640643 | |
NMWCVZCSJHJYFW-UHFFFAOYSA-M | |
sodium;3,5-dichloro-2-hydroxybenzenesulfonate | |
3085077 | |
99% |
>300°C | |
MFCD00009798 | |
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt | |
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+] | |
265.036 | |
265.05 | |
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 259-416-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : 3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.