missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

43.52 € - 191.96 €

Chemical Identifiers

CAS 23378-88-3
Molecular Formula C6H3Cl3O3S
Molecular Weight (g/mol) 261.50
MDL Number MFCD00007432
InChI Key KXFQRJNVGBIDHA-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
PubChem CID 90078
IUPAC Name 3,5-dichloro-2-hydroxybenzenesulfonyl chloride
SMILES OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388635
View Documents
Thermo Scientific Alfa Aesar
L12147.09
10 g
43.52 €
10g
Please to purchase this item. Need a web account? Register with us today!
11398635
View Documents
Thermo Scientific Alfa Aesar
L12147.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5?-nucleotidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5′-nucleotidase.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
Specifications

Chemical Identifiers

23378-88-3
261.50
KXFQRJNVGBIDHA-UHFFFAOYSA-N
90078
OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O
C6H3Cl3O3S
MFCD00007432
3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
3,5-dichloro-2-hydroxybenzenesulfonyl chloride

Specifications

23378-88-3
C6H3Cl3O3S
UN3261
3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O
261.50
261.51
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride
78°C to 80°C
MFCD00007432
2940993
KXFQRJNVGBIDHA-UHFFFAOYSA-N
3,5-dichloro-2-hydroxybenzenesulfonyl chloride
90078
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.