missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11481638
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-43-7
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007774
InChI Key UQRLKWGPEVNVHT-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline
PubChem CID 12281
ChEBI CHEBI:19904
IUPAC Name 3,5-dichloroaniline
SMILES C1=C(C=C(C=C1Cl)Cl)N

Specifications

Melting Point 49°C to 53°C
Boiling Point 259°C to 260°C (741 mmHg)
Flash Point 133°C (271°F)
Quantity 1000 g
UN Number UN3442
Beilstein 636492
Formula Weight 162.02
Percent Purity 98%
Chemical Name or Material 3,5-Dichloroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'