missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11450933
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10203-08-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003352
InChI Key CASRSOJWLARCRX-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dichloro, 3,5-dichloro-benzaldehyde, unii-m74yu7se79, rarechem aq a2 0002, pubchem3068, 3,5-dichlorobezaldehyde, 3,5-dichlorobenaldehyde, 3?5-dichlorobenzaldehyde, benzaldehyde,5-dichloro, acmc-1bxq6
PubChem CID 35746
IUPAC Name 3,5-dichlorobenzaldehyde
SMILES C1=C(C=C(C=C1Cl)Cl)C=O

Specifications

Melting Point 60°C to 66°C
Boiling Point 235°C to 240°C
Quantity 1 g
UN Number UN1759
Beilstein 2040553
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'