missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorobenzaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.
Solubility
Insoluble in water.
Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.
Solubility
Insoluble in water.
Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | 60°C to 66°C |
Boiling Point | 235°C to 240°C |
Quantity | 25 g |
UN Number | UN1759 |
Beilstein | 2040553 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 175.01 |
Percent Purity | 97% |
Chemical Name or Material | 3,5-Dichlorobenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'