missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenol, 98%, Thermo Scientific Chemicals

56.08 € - 313.56 €

Chemical Identifiers

CAS 591-35-5
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 162.997
MDL Number MFCD00002259
InChI Key VPOMSPZBQMDLTM-UHFFFAOYSA-N
Synonym phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
PubChem CID 11571
IUPAC Name 3,5-dichlorophenol
SMILES C1=C(C=C(C=C1Cl)Cl)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11309529
View Documents
Thermo Scientific Alfa Aesar
A15328.06
5 g
56.08 €
5g
Please to purchase this item. Need a web account? Register with us today!
11319529
View Documents
Thermo Scientific Alfa Aesar
A15328.14
25 g
186.93 €
25g
Please to purchase this item. Need a web account? Register with us today!
11488226
View Documents
Thermo Scientific Alfa Aesar
A15328.18
50 g
313.56 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3,5-Dichlorophenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenol is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water. Very soluble in ethanol, ethyl ether and petroleum ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

591-35-5
162.997
VPOMSPZBQMDLTM-UHFFFAOYSA-N
11571
C1=C(C=C(C=C1Cl)Cl)O
C6H4Cl2O
MFCD00002259
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
3,5-dichlorophenol

Specifications

591-35-5
233°C
C6H4Cl2O
UN2020
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
VPOMSPZBQMDLTM-UHFFFAOYSA-N
3,5-dichlorophenol
11571
98%
63°C to 68°C
Phenol-like
MFCD00002259
1906458
Slightly soluble in water. Very soluble in ethanol,ethyl ether and petroleum ether.
C1=C(C=C(C=C1Cl)Cl)O
162.997
163
3,5-Dichlorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID

missing translation for 'einecsNumber' : 209-714-9

missing translation for 'rtecsNumber' : SK8820000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.