Learn More
3,5-Dichlorophenylacetic acid, 95%, Thermo Scientific Chemicals
28.90 € - 325.00 €
Chemical Identifiers
CAS | 51719-65-4 |
---|---|
Molecular Formula | C8H6Cl2O2 |
Molecular Weight (g/mol) | 205.034 |
MDL Number | MFCD01764671 |
InChI Key | RERINLRFXYGZEE-UHFFFAOYSA-N |
Synonym | 2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid |
PubChem CID | 6452487 |
IUPAC Name | 2-(3,5-dichlorophenyl)acetic acid |
SMILES | C1=C(C=C(C=C1Cl)Cl)CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15409928
|
Thermo Scientific Alfa Aesar
H63293.MD |
250 mg | N/A | ||||||
15489918
|
Thermo Scientific Alfa Aesar
H63293.03 |
1 g |
84.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15499918
|
Thermo Scientific Alfa Aesar
H63293.06 |
5 g |
325.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51719-65-4 | |
205.034 | |
RERINLRFXYGZEE-UHFFFAOYSA-N | |
6452487 | |
C1=C(C=C(C=C1Cl)Cl)CC(=O)O |
C8H6Cl2O2 | |
MFCD01764671 | |
2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid | |
2-(3,5-dichlorophenyl)acetic acid |
Specifications
51719-65-4 | |
MFCD01764671 | |
RERINLRFXYGZEE-UHFFFAOYSA-N | |
2-(3,5-dichlorophenyl)acetic acid | |
6452487 | |
95% | |
3,5-Dichlorophenylacetic acid |
C8H6Cl2O2 | |
2-3,5-dichlorophenyl acetic acid, 3,5-dichlorophenylacetic acid, benzeneacetic acid, 3,5-dichloro, 3,5-dichlorobenzeneacetic acid, 3,5-dichlorophenyl acetic acid, 3,5-dichloro-phenyl-acetic acid, 2-3,5-dichlorophenyl-acetic acid, 3,5-dichlorophenylaceticacid, pubchem24116, 3,5-dichlorphenylacetic acid | |
C1=C(C=C(C=C1Cl)Cl)CC(=O)O | |
205.034 | |
205.03 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only