missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',5'-Difluoro-4'-methoxyacetophenone, 97%, Thermo Scientific™
90.70 € - 303.00 €
Chemical Identifiers
CAS | 170570-79-3 |
---|---|
Molecular Formula | C9H8F2O2 |
Molecular Weight (g/mol) | 186.158 |
MDL Number | MFCD00236243 |
InChI Key | OUJZNFJFMAKGMS-UHFFFAOYSA-N |
Synonym | 1-3,5-difluoro-4-methoxyphenyl ethanone, 3',5'-difluoro-4'-methoxyacetophenone, 3,5-difluoro-4-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 1-3,5-difluoro-4-methoxy-phenyl ethanone, 4-acetyl-2,6-difluoroanisole, 3.5-difluoro-4-methoxyacetophenone, 1-acetyl-3,5-difluoro-4-methoxybenzene, 3',5'-difluoro-4-difluoro-4-methoxyacetophenone-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-2,6-difluoroanisole |
PubChem CID | 2736983 |
IUPAC Name | 1-(3,5-difluoro-4-methoxyphenyl)ethanone |
SMILES | CC(=O)C1=CC(=C(C(=C1)F)OC)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3′,5′-Difluoro-4′-methoxyacetophenone, is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
170570-79-3 | |
186.158 | |
OUJZNFJFMAKGMS-UHFFFAOYSA-N | |
2736983 | |
CC(=O)C1=CC(=C(C(=C1)F)OC)F |
C9H8F2O2 | |
MFCD00236243 | |
1-3,5-difluoro-4-methoxyphenyl ethanone, 3',5'-difluoro-4'-methoxyacetophenone, 3,5-difluoro-4-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 1-3,5-difluoro-4-methoxy-phenyl ethanone, 4-acetyl-2,6-difluoroanisole, 3.5-difluoro-4-methoxyacetophenone, 1-acetyl-3,5-difluoro-4-methoxybenzene, 3',5'-difluoro-4-difluoro-4-methoxyacetophenone-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-2,6-difluoroanisole | |
1-(3,5-difluoro-4-methoxyphenyl)ethanone |
Specifications
170570-79-3 | |
MFCD00236243 | |
Insoluble in water. | |
CC(=O)C1=CC(=C(C(=C1)F)OC)F | |
186.158 | |
186.15 | |
3',5'-Difluoro-4'-methoxyacetophenone |
C9H8F2O2 | |
1-3,5-difluoro-4-methoxyphenyl ethanone, 3',5'-difluoro-4'-methoxyacetophenone, 3,5-difluoro-4-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 1-3,5-difluoro-4-methoxy-phenyl ethanone, 4-acetyl-2,6-difluoroanisole, 3.5-difluoro-4-methoxyacetophenone, 1-acetyl-3,5-difluoro-4-methoxybenzene, 3',5'-difluoro-4-difluoro-4-methoxyacetophenone-methoxyacetophenone, 1-3,5-difluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-2,6-difluoroanisole | |
OUJZNFJFMAKGMS-UHFFFAOYSA-N | |
1-(3,5-difluoro-4-methoxyphenyl)ethanone | |
2736983 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures